missing translation for 'onlineSavingsMsg'
Learn More

Thiazovivin, Tocris Bioscience™

Artikelnummer. 18045212
Click to view available options
Menge:
10 mg
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

ROCK inhibitor ; improves the efficiency of fibroblast reprogramming and induction of iPSCs

Thiazovivin is a selective, cell-permeable Rho-associated coiled-coil containing protein kinase (ROCK) inhibitor (IC50 = 0.5 μM). Thiazovivin enhances the efficiency of fibroblast reprogramming to generate induced pluripotent stem cells (iPSCs) when used in combination with SB 431542 (Cat. No. 1614) and PD 0325901 (Cat. No. 4192). Thiazovivin improves the survival of human embryonic stem cells (hESCs) upon trypsinization and increases cell adhesion through the regulation of E-cadherin and significantly improves direct reprogramming efficiency. In combination with Valproic acid, sodium salt (Cat. No. 2815), Purmorphamine (Cat. No. 4551) and A 83-01 (Cat. No. 2939), Thiazovivin can be used to directly reprogram mouse embryonic fibroblasts into chemically induced neural stem cells.

Spezifikation

Chemischer Name oder Material Thiazovivin
CAS 1226056-71-8
Summenformel C15H13N5OS
Menge 10 mg
Synonym thiazovivin, n-benzyl-2-pyrimidin-4-ylamino thiazole-4-carboxamide, n-benzyl-2-pyrimidin-4-ylamino-1,3-thiazole-4-carboxamide, n-benzyl-2-pyrimidin-4-ylamino-thiazole-4-carboxamide, n-phenylmethyl-2-4-pyrimidinylamino-4-thiazolecarboxamide, pubchem24336, n-benzyl-2-pyrimidin-4-yl amino thiazole-4-carboxamide, benzyl-2-pyrimidin-4-ylamino thiazole-4-carboxamide, n-benzyl-2-pyrimidin-4-yl amino-1,3-thiazole-4-carboxamide, 4-thiazolecarboxamide, n-phenylmethyl-2-4-pyrimidinylamino
InChI-Schlüssel DOBKQCZBPPCLEG-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)CNC(=O)C2=CSC(=N2)NC3=NC=NC=C3
IUPAC-Name N-benzyl-2-(pyrimidin-4-ylamino)-1,3-thiazole-4-carboxamide
Molekulargewicht (g/mol) 311.363
PubChem CID 46209426
Formelmasse 311.36
Reinheit (%) 98%
Mehr anzeigen Weniger anzeigen
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'