missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBP Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 22006-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
This antibody recognize human TBP with MW 40 kDa and mouse/rat Tbp with MW 34 kDa.
RICH1 is a GTPase-activating protein (GAP). GAPs stimulate the intrinsic GTP hydrolysis of small G proteins, such as RHOA, RAC1, and CDC42.Spezifikation
TBP | |
Polyclonal | |
Unconjugated | |
TBP | |
GTF2D, GTF2D1, SCA17, TATA binding factor, TATA box binding protein, TATA box factor, TATA sequence binding protein, TBP, TF2D, TFIID | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
21374, 6908 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.32 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P20226, P29037 | |
TBP | |
TBP Fusion Protein Ag12383 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |