Learn More
Berkshire Super Polx™ SWSE Irradiated
Super Polx™ SWSE Irradiated is a sealed edge cleanroom laundered wiper recommended for ISO Class 3 and above environments.
Marke: Berkshire SPSWSE121230IR
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
- 100% continuous filament double knitted polyester
- Ultrasonically sealed edge
- Laundered and packaged in an ISO Class 4 (Class 10) cleanroom
- Lot tracing on each pack
- Economical
- Strong, tear and abrasion resistant
- Very good absorbency
- Low particles and fibers
- Soft hand and edge for sensitive surfaces
Spezifikation
Surface Wiper | |
30 cm | |
30 cm | |
Not Autoclavable | |
1-ply |
12 in. | |
12 in. | |
Polyester | |
Single-use | |
Sterile |