Learn More
Cole-Parmer Stepless Ear Clamps
Unique stepless design provides a 360° leak-free seal
Marke:  Cole-Parmer 50117-32
Artikelnummer. 30053818
  • 14.55 CHF / 10 Stück
Nur noch null auf Lager
Verfügbar ab: 28-08-2025
Beschreibung
Tongue-in-groove design e in., erts uniform pressure. Dimple provides increased clamping force. Fast and easy installation. Degreased to minimize contamination. Lightweight, narrow band design
Zertifikate
Aktionen
Aktionen
Berichtigung von Produktinhalten
Name des Produkts
Cole-Parmer Stepless Ear Clamps > 10 Stepless Ear Clamp 12 to 14.5mm
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.