Learn More
Blanco™ Stainless Steel Bath Tub Trolley
The bain-marie Stainless Steel Bath Tub Trolley for use with laboratory or industry applications to heat materials and substances gently, or to keep warm over a period of time at a constant temperature. The warming trays can be heated wet or dry using the adjustable thermostat.
Marke:  Blanco™ 572152
Artikelnummer. 11928017
  • 5675.00 CHF / 1 Stück
Verfügbar ab: 28-02-2025
Sie sich ein, um den Lagerbestand zu sehen
Entdecken Sie weitere Sonderangebote
Beschreibung
  • Can be heated wet or dry using the adjustable thermostat.
Spezifikation
Spezifikation
In accordance with DIN 18867, Section 7; CE conformity
Wasserbad
Stainless steel
0.7 kW
20 in.
36.7 in.
8.5 in.
54 lb.
220/240 V, 50/60 Hz
30 °C bis 95 °C
1 individual well for 1 x GN 1/1-200 or subdivision
SAW1
750 mm
508 mm
933 mm
215 mm
24.5 kg