missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SERCA3 ATPase Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA5111764
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Reconstitute antibody with 50 μL of double distilled water (DDW). The reconstituted solution can be stored at 4 degrees C for up to 1 week. For longer periods, small aliquots should be stored at -20 degrees C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min). Peptide sequence is: (C)DEALKYL SRHHVD Homology: Mouse - 11/13 amino acid residues identical; human - 10/13 amino acid residues identical.
ATP dependent calcium pumps are responsible in part for the maintenance of low cytoplasmic free calcium concentrations. The ATP pumps that reside in intracellular organelles are encoded by a family of structurally related enzymes, termed the sarcoplasmic or endoplasmic reticulum calcium ATPases (SERCA). The SERCA2 gene is subject to tissue dependent processing which is responsible for the generation of SERCA2a muscle-specific isoform expressed in type I (slow) skeletal, cardiac and smooth muscle and the SERCA2b isoform expressed in all cell types. SERCA3 has been found to co-express with SERCA2b in platelets, mast cells, lymphoid cells, and epithelial cells. Due to this pattern of expression it is referred to as the non-muscle form of SERCA.Spezifikation
SERCA3 ATPase | |
Polyclonal | |
Unconjugated | |
Atp2a3 | |
Atp2a3; Calcium pump 3; Sarcoplasmic/endoplasmic reticulum calcium ATPase 3; SERCA3; SERCA3b | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
25391, 53313 | |
-20°C | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Western Blot | |
0.8 mg/mL | |
PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
P18596, Q64518 | |
Atp2a3 | |
Peptide corresponding to amino acid residues 981-993 of rat SERCA3 (Cytoplasmic, C-terminus) | |
50 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur