Learn More
Novus Biologicals™ Selenoprotein W Overexpression Lysate
Marke:  Novus Biologicals™
Artikelnummer. 18144750
  • 820.00 CHF / 0.10 Milligramm
Verfügbar ab: 10-01-2025
Sie sich ein, um den Lagerbestand zu sehen
Entdecken Sie weitere Sonderangebote
Beschreibung
The Selenoprotein W Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
Spezifikation
Spezifikation
The lysate was created in HEK293T cells, using plasmid ID RC215316 and based on accession number NP_3. The protein contains a C-terminal DDK tag.
0.1 mg
Protein
-80°C. Avoid freeze-thaw cycles.
Western Blot
RIPA buffer
Homo sapiens Selenoprotein W, mRNA.
Native
Sonderangebote
Nur für Forschungszwecke