Learn More
Novus Biologicals™ RUNDC2B Overexpression Lysate
Marke: Novus Biologicals™ NBL1-15622
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The RUNDC2B Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.Spezifikation
The lysate was created in HEK293T cells, using plasmid ID RC28352 and based on accession number NM_112391. The protein contains a C-terminal DDK tag. | |
0.1 mg | |
Protein | |
-80°C. Avoid freeze-thaw cycles. |
Western Blot | |
RIPA buffer | |
Homo sapiens RUN domain containing 2B (RUNDC2B), mRNA. | |
Native |
Nur für Forschungszwecke