Learn More
R&D Systems™ Recombinant Viral B8R Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Marke:  R&D Systems™ 8225-BR-025/CF
 Lernen Sie weitere Varianten dieses
Artikelnummer. 18757377
  • 402.00 CHF / 25 Mikrogramm
Verfügbar ab: 15-01-2025
Sie sich ein, um den Lagerbestand zu sehen
Entdecken Sie weitere Sonderangebote
Beschreibung
The Recombinant Viral B8R Protein is derived from CHO. The Recombinant Viral B8R Protein has been validated for the following applications: Bioactivity.
Bestellinformation
The product is shipped at ambient temperature, upon receipt, store immediately
Spezifikation
Spezifikation
Lyophilized from a 0.2μm filtered solution in PBS
Chinese Hamster Ovary cell line,CHO-derived viral B8R protein Lys18-Ser272,with a C-terminal 6-His tag
Unconjugated
25 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.