Learn More
R&D Systems™ Recombinant Rat LIX Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
534.00 CHF
Spezifikation
Gen-ID (Entrez) | 60665 |
---|---|
Molekulargewicht | 10.1 kDa |
Name | LIX |
Menge | 25 μg |
Quelle | E. coli-derived rat LIX protein Ala38-Arg125 & Ala38-Lys117 |
Produktcode | Marke | Zusammensetzung | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Zusammensetzung | Preis | Menge & Verfügbarkeit | |||||
18345627
|
R&D Systems™
543-RL-025 |
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. |
534.00 CHF
25 Mikrogramm |
Verfügbar ab: 22-01-2025
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
18319467
|
R&D Systems™
543-RL-025/CF |
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. |
534.00 CHF
25 Mikrogramm |
Verfügbar ab: 22-01-2025
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
The Recombinant Rat LIX Protein is derived from E. coli. The Recombinant Rat LIX Protein has been validated for the following applications: Bioactivity.Spezifikation
60665 | |
LIX | |
E. coli-derived rat LIX protein Ala38-Arg125 & Ala38-Lys117 | |
Unconjugated | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
10.1 kDa | |
25 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur