missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant other Protein-A/G/L Protein
A cDNA sequence encoding the Protein-A/G/L was constructed and used to recombinantly synthesize the protein.
143.00 CHF - 1880.00 CHF
Spezifikation
Name | Protein-A/G/L Protein |
---|---|
Kennzeichnung | Research Use Only |
Endotoxin-Konzentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Produkttyp | Recombinant Protein |
Kreuzreaktivität | Other |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15992699
|
enQuireBio™
QP13165-1mg |
1 mg |
143.00 CHF
1 Milligramm |
Verfügbar ab: 11-12-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15982699
|
enQuireBio™
QP13165-10mg |
10 mg |
384.00 CHF
10 Milligramm |
Verfügbar ab: 11-12-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15972699
|
enQuireBio™
QP13165-100mg |
100 mg |
1880.00 CHF
100 Milligramm |
Verfügbar ab: 11-12-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Spezifikation
Protein-A/G/L Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Other | |
Untagged | |
Protein- A/G/L was lyophilized without any additives. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
NAAQHDEAQQ NAFYQVLNMP NLNADQRNGF IQSLKDDPSQ SANVLGEAQK LNDSQAPKAD AQQNNFNKDQ QSAFYEILNM PNLNEAQRNG FIQSLKDDPS QSTNVLGEAK KLNESQAPKA DNNFNKEQQN AFYEILNMPN LNEEQRNGFI QSLKDDPSQS ANLLSEAKKL NESQAPKADN KFNKEQQNAF YEILHLPNLN EEQRNGFIQS LKDDPSQSAN LLAEAKKLND AQAPKADNKF NKEQQNAFYE ILHLPNLTEE QRNGFIQSLK DDPSVSKEIL AEAKKLNDAQ APKEEDSLEG SGSGTYKLIL NGKTLKGETT TEAVDAATAE KVFKQYANDN GVDGEWTYDD ATKTFTVTEK PEVIDASELT PAVTTYKLVI NGKTLKGETT TKAVDAETAE KAFKQYANDN GVDGVWTYDD ATKTFTVTEE PRARPGSGSG KEETPETPET DSEEEVTIKA NLIFANGSTQ TAEFKGTFEK ATSEAYAYAD TLKKDNGEYT VDVADKGYTL NIKFAGKEKT PEEPKEEVTI KANLIYADGK TQTAEFKGTF EEATAEAYRY ADALKKDNGE YTVDVADKGY TLNIKFAGKE KTPEEPKEEV TIKANLIYAD GKTQTAEFKG TFEEATAEAY RYADLLAKEN GKYTVDVADK GYTLNIKFAG KEKTPEEPKE EVTIKANLIY ADGKTQTAEF KGTFAEATAE AYRYADLLAK ENGKYTADLE DGGYTINIRF AGKKVDEKPE EKEQVTIKEN IYFEDGTVQT ATFKGTFAEA TAEAYRYADL LSKEHGKYTA DLEDGGYTIN IRFAG | |
Greater than 95.0% as determined by: (a) Analysis by SEC-HPLC. (b) Analysis by SDS-PAGE. |