missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse NRP1 Protein
A cDNA sequence encoding the NRP1 was constructed and used to recombinantly synthesize the protein.
131.00 CHF - 1735.00 CHF
Spezifikation
Name | NRP1 Protein |
---|---|
Kennzeichnung | Research Use Only |
Endotoxin-Konzentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Produkttyp | Recombinant Protein |
Kreuzreaktivität | Mouse |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15971839
|
enQuireBio™
QP12883-1ug |
1 μg |
131.00 CHF
1 Mikrogramm |
Verfügbar ab: 29-10-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15991839
|
enQuireBio™
QP12883-5ug |
5 μg |
203.00 CHF
5 Mikrogramm |
Verfügbar ab: 29-10-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15981839
|
enQuireBio™
QP12883-50ug |
50 μg |
1735.00 CHF
50 Mikrogramm |
Verfügbar ab: 29-10-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Spezifikation
NRP1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Mouse | |
His | |
NRP1 protein solution (0.25 mg/ml) containing Phosphate Buffered Saline (pH 7.4) and 20% glycerol. |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
FRSDKCGGTI KIENPGYLTS PGYPHSYHPS EKCEWLIQAP EPYQRIMINF NPHFDLEDRD CKYDYVEVID GENEGGRLWG KFCGKIAPSP VVSSGPFLFI KFVSDYETHG AGFSIRYEIF KRGPECSQNY TAPTGVIKSP GFPEKYPNSL ECTYIIFAPK MSEIILEFES FDLEQDSNPP GGMFCRYDRL EIWDGFPEVG PHIGRYCGQK TPGRIRSSSG VLSMVFYTDS AIAKEGFSAN YSVLQSSISE DFKCMEALGM ESGEIHSDQITASSQYGTNW SVERSRLNYP ENGWTPGEDS YKEWIQVDLG LLRFVTAVGT QGAISKETKK KYYVKTYRVD ISSNGEDWIS LKEGNKAIIF QGNTNPTDVV LGVFSKPLIT RFVRIKPVSW ETGISMRFEV YGCKITDYPC SGMLGMVSGL ISDSQITASN QADRNWMPEN IRLVTSRTGW ALPPSPHPYT NEWLQVDLGD EKIVRGVIIQ GGKHRENKVF MRKFKIAYSN NGSDWKTIMD DSKRKAKSFE GNNNYDTPEL RTFSPLSTRF IRIYPERATH SGLGLRMELL GCEVEAPTAG PTTPNGNPVD ECDDDQANCH SGTGDDFQLT GGTTVLATEK PTIIDSTIQS EFPTYGFNCE FGWGSHKTFC HWEHDSHAQL RWSVLTSKTG PIQDHTGDGN FIYSQADENQ KGKVARLVSP VVYSQSSAHC MTFWYHMSGS HVGTLRVKLR YQKPEEYDQL VWMVVGHQGD HWKEGRVLLH KSLKLYQVIF EGEIGKGNLG GIAVDDISIN NHISQEDCAK PTDLDKKNTE IKIDETGSTP GYEGEGEGDK NISRKPGNVL KTLDPLEHHH HHH | |
Greater than 90.0% as determined by SDS-PAGE. |