Learn More
R&D Systems™ Recombinant Mouse CXCL16 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Marke:  R&D Systems™ 503-CX-025/CF
 Lernen Sie weitere Varianten dieses
Artikelnummer. 18166520
  • 381.00 CHF / 25 Mikrogramm
Verfügbar ab: 03-02-2025
Sie sich ein, um den Lagerbestand zu sehen
Entdecken Sie weitere Sonderangebote
Beschreibung
The Recombinant Mouse CXCL16 Protein is derived from E. coli. The Recombinant Mouse CXCL16 Protein has been validated for the following applications: Bioactivity.
Spezifikation
Spezifikation
Lyophilized from a 0.2μm filtered solution in PBS.
9.9 kDa
25 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant
66102
CXCL16 Chemokine Domain
E. coli-derived mouse CXCL16 protein Asn27-Pro114
Unconjugated
>97%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.