missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse CLEC4F/CLECSF13 Protein
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Marke: R&D Systems™ 2784-CL-050
410.36 CHF gültig bis 2024-12-16
Benutzen Sie den Promotions-Code "21630" um Ihren Promotions-Preis zu erhalten.
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The Recombinant Mouse CLEC4F/CLECSF13 Protein is derived from NS0. The Recombinant Mouse CLEC4F/CLECSF13 Protein has been validated for the following applications: Binding Activity.Spezifikation
Lyophilized from a 0.2μm filtered solution in PBS. | |
55 kDa | |
50 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
51811 | |
CLEC4F/CLECSF13 | |
Mouse myeloma cell line,NS0-derived mouse CLEC4F/CLECSF13 protein Ala65-Gly548,with an N-terminal 9-His tag | |
Unconjugated | |
85%, by SDS-PAGE under reducing conditions and visualized by silver stain. |