missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse Angiopoietin-like 4 Protein
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Inhibition Activity
Marke: R&D Systems™ 4880-AN-050
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The Recombinant Mouse Angiopoietin-like 4 Protein is derived from CHO. The Recombinant Mouse Angiopoietin-like 4 Protein has been validated for the following applications: Inhibition Activity.Spezifikation
Lyophilized from a 0.2μm filtered solution in PBS and NaCl. | |
44.2 kDa | |
50 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
57875 | |
Angiopoietin-like Protein 4/ANGPTL4 | |
Chinese Hamster Ovary cell line,CHO-derived mouse Angiopoietin-like Protein 4/ANGPTL4 protein Gln24-Ser410,with a C-terminal 6-His tag | |
Oligomer | |
Recombinant |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur