Learn More
R&D Systems™ Recombinant Human Syntaxin 4 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Marke:  R&D Systems™ 7894-ST-050
Artikelnummer. 18128350
  • 538.00 CHF / 50 Mikrogramm
Verfügbar ab: 04-02-2025
Sie sich ein, um den Lagerbestand zu sehen
Entdecken Sie weitere Sonderangebote
Beschreibung
The Recombinant Human Syntaxin 4 Protein is derived from E. coli. The Recombinant Human Syntaxin 4 Protein has been validated for the following applications: Bioactivity.
Spezifikation
Spezifikation
Lyophilized from a 0.2μm filtered solution in PBS and TCEP.
22.2 kDa
50μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant
6810
Syntaxin 4
E. coli-derived human Syntaxin 4 protein Met1-Glu183,with a C-terminal 6-His tag
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.