missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Plexin B3 Protein

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Marke:  R&D Systems™ 4958-PC-050

444.91 CHF gültig bis 2024-12-16
Benutzen Sie den Promotions-Code "21630" um Ihren Promotions-Preis zu erhalten.


Product Code. 18136930

  • 569.00 CHF / 50 Mikrogramm
Verfügbar ab: 19-12-2024
Sie sich ein, um den Lagerbestand zu sehen
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

The Recombinant Human Plexin B3 Protein is derived from NS0. The Recombinant Human Plexin B3 Protein has been validated for the following applications: Binding Activity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
129.6 kDa
50μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant
5365
Plexin B3
Mouse myeloma cell line,NS0-derived human Plexin B3 protein His45-Gln1255 (Glu1156Asp),with a C-terminal 6-His tag
Unconjugated
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers