missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human pGH 20kDa Protein
A cDNA sequence encoding the pGH 20kDa was constructed and used to recombinantly synthesize the protein.
131.00 CHF - 3040.00 CHF
Spezifikation
Name | pGH 20kDa Protein |
---|---|
Kennzeichnung | Research Use Only |
Endotoxin-Konzentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Produkttyp | Recombinant Protein |
Kreuzreaktivität | Human |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15962289
|
enQuireBio™
QP13029-10ug |
10 μg |
131.00 CHF
10 Mikrogramm |
Verfügbar ab: 11-12-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15982289
|
enQuireBio™
QP13029-50ug |
50 μg |
203.00 CHF
50 Mikrogramm |
Verfügbar ab: 11-12-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15972289
|
enQuireBio™
QP13029-1mg |
1 mg |
3040.00 CHF
1 Milligramm |
Verfügbar ab: 11-12-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Spezifikation
pGH 20kDa Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
Greater than 98.0% as determined by:(a) Analysis by SEC-HPLC. (b) Analysis by SDS-PAGE. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
pGH 20kDa was lyophilized from a concentrated (1 mg/ml) solution with0.0045mM NaHCO3 previously adjusted pH 11. |