Learn More
enQuireBio™ Recombinant Human PFDN2 Protein
A cDNA sequence encoding the PFDN2 was constructed and used to recombinantly synthesize the protein.
141.00 CHF - 8085.00 CHF
Spezifikation
Name | PFDN2 Protein |
---|---|
Kennzeichnung | Research Use Only |
Endotoxin-Konzentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Produkttyp | Recombinant Protein |
Kreuzreaktivität | Human |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15992249
|
enQuireBio™
QP13015-2ug |
2 μg |
141.00 CHF
2 Mikrogramm |
Verfügbar ab: 18-02-2025
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15972249
|
enQuireBio™
QP13015-10ug |
10 μg |
219.00 CHF
10 Mikrogramm |
Verfügbar ab: 18-02-2025
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15982249
|
enQuireBio™
QP13015-1mg |
1 mg |
8085.00 CHF
1 Milligramm |
Verfügbar ab: 18-02-2025
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Spezifikation
PFDN2 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The PFDN2 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol, 1mM DTT and 50mM NaCl. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MAENSGRAGK SSGSGAGKGA VSAEQVIAGF NRLRQEQRGL ASKAAELEME LNEHSLVIDT LKEVDETRKC YRMVGGVLVE RTVKEVLPAL ENNKEQIQKI IETLTQQLQA KGKELNEFRE KHNIRLMGED EKPAAKENSE GAGAKASSAG VLVS | |
Greater than 90.0% as determined by SDS-PAGE. |