missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PAPP A Protein
A cDNA sequence encoding the PAPP A was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12957-2ug
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Spezifikation
PAPP A Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MRLWSWVLHL GLLSAALGCG LAERPRRARR DPRAGRPPRP AAGPATCATR AARGRRASPP PPPPPGGAWE AVRVPRRRQQ REARGATEEP SPPSRALYFS GRGEQLRLRA DLELPRDAFT LQVWLRAEGG QRSPAVITGL YDKCSYISRD RGWVVGIHTI SDQDNKDPRY FFSLKTDRAR QVTTINAHRS YLPGQWVYLA ATYDGQFMKL YVNGAQVATS GEQVGGIFSP LTQKCKVLML GGSALNHNYR GYIEHFSLWK VARTQREILS DMETHGAHTA LPQLLLQENW DNVKHAWSPM KDGSSPKVEF SNAHGFLLDT SLEPPLCGQT LCDNTEVIAS YNQLSSFRQP KVVRYRVVNL YEDDHKNPTV TREQVDFQHH QLAEAFKQYN ISWELDVLEV SNSSLRRRLI LANCDISKIG DENCDPECNH TLTGHDGGDC RHLRHPAFVK KQHNGVCDMD CNYERFNFDG GECCDPEITN VTQTCFDPDS PHRAYLDVNE LKNILKLDGS THLNIFFAKS SEEELAGVAT WPWDKEALMH LGGIVLNPSF YGMPGHTHTM IHEIGHSLGL YHVFRGISEI QSCSDPCMET EPSFETGDLC NDTNPAPKHK SCGDPGPGND TCGFHSFFNT PYNNFMSYAD DDCTDSFTPN QVARMHCYLD LVYQGWQPSR KPAPVALAPQ VLGHTTDSVT LEWFPPIDGH FFERELGSAC HLCLEGRILV QYASNASSPM PCSPSGHWSP REAEGHPDVE QPCKSSVRTW SPNSAVNPHT VPPACPEPQG CYLELEFLYP LVPESLTIWV TFVSTDWDSS GAVNDIKLLA VSGKNISLGP QNVFCDVPLT IRLWDVGEEV YGIQIYTLDE HLEIDAAMLT STADTPLCLQ CKPLKYKVVR DPPLQMDVAS ILHLNRKFVD MDLNLGSVYQ YWVITISGTE ESEPSPAVTY IHGSGYCGDG IIQKDQGEQC DDMNKINGDG CSLFCRQEVS FNCIDEPSRC YFHDGDGVCE EFEQKTSIKD CGVYTPQGFL DQWASNASVS HQDQQCPGWV IIGQPAASQV CRTKVIDLSE GISQHAWYPC TISYPYSQLA QTTFWLRAYF SQPMVAAAVI VHLVTDGTYY GDQKQETISV QLLDTKDQSH DLGLHVLSCR NNPLIIPVVH DLSQPFYHSQ AVRVSFSSPL VAISGVALRS FDNFDPVTLS SCQRGETYSP AEQSCVHFAC EKTDCPELAV ENASLNCSSS DRYHGAQCTV SCRTGYVLQI RRDDELIKSQ TGPSVTVTCT EGKWNKQVAC EPVDCSIPDH HQVYAASFSC PEGTTFGSQC SFQCRHPAQL KGNNSLLTCM EDGLWSFPEA LCELMCLAPP PVPNADLQTA RCRENKHKVG SFCKYKCKPG YHVPGSSRKS KKRAFKTQCT QDGSWQEGAC VPVTCDPPPP KFHGLYQCTN GFQFNSECRI KCEDSDASQG LGSNVIHCRK DGTWNGSFHV CQEMQGQCSV PNELNSNLKL QCPDGYAIGS ECATSCLDHN SESIILPMNV TVRDIPHWLN PTRVERVVCT AGLKWYPHPA LIHCVKGCEP FMGDNYCDAI NNRAFCNYDG GDCCTSTVKT KKVTPFPMSC DLQGDCACRD PQAQEHSRKD LRGYSHG | |
Greater than 95.0% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
PAPP-A protein is supplied in 1x PBS and 50% glycerol. |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur