missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NUP62 Protein
A cDNA sequence encoding the NUP62 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12909-20ug
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Spezifikation
NUP62 Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
MSGFNFGGTG APTGGFTFGT AKTATTTPAT GFSFSTSGTG GFNFGAPFQP ATSTPSTGLF SLATQTPATQ TTGFTFGTAT LASGGTGFSL GIGASKLNLS NTAATPAMAN PSGFGLGSSN LTNAISSTVT SSQGTAPTGF VFGPSTTSVA PATTSGGFSF TGGSTAQPSG FNIGSAGNSA QPTAPATLPF TPATPAATTA GATQPAAPTP TATITSTGPS LFASIATAPT SSATTGLSLC TPVTTAGAPT AGTQGFSLKA PGAASGTSTT TSTAATATAT TTSSSSTTGF ALNLKPLAPA GIPSNTAAAV TAPPGPGAAA GAAASSAMTY AQLESLINKW SLELEDQERH FLQQATQVNA WDRTLIENGE KITSLHREVE KVKLDQKRLD QELDFILSQQ KELEDLLSPL EELVKEQSGT IYLQHADEER EKTYKLAENI DAQLKRMAQD LKDIIEHLNT SGAPADTSDP LQQICKILNA HMDSLQWIDQ NSALLQRKVE EVTKVCEGRR KEQERSFRIT FD | |
Greater than 80% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
NUP62 is supplied in 20mM HEPES buffer pH 7.6, 250mM NaCl and 20% glycerol. |