missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NOB1 Protein
A cDNA sequence encoding the NOB1 was constructed and used to recombinantly synthesize the protein.
131.00 CHF - 3905.00 CHF
Spezifikation
Name | NOB1 Protein |
---|---|
Kennzeichnung | Research Use Only |
Endotoxin-Konzentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Produkttyp | Recombinant Protein |
Kreuzreaktivität | Human |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15991779
|
enQuireBio™
QP12863-5ug |
5 μg |
131.00 CHF
5 Mikrogramm |
Verfügbar ab: 11-12-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15981779
|
enQuireBio™
QP12863-20ug |
20 μg |
203.00 CHF
20 Mikrogramm |
Verfügbar ab: 11-12-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15971779
|
enQuireBio™
QP12863-1mg |
1 mg |
3905.00 CHF
1 Milligramm |
Verfügbar ab: 11-12-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Spezifikation
NOB1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The NOB1 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAPVEHV VADAGAFLRH AALQDIGKNI YTIREVVTEI RDKATRRRLA VLPYELRFKE PLPEYVRLVT EFSKKTGDYP SLSATDIQVL ALTYQLEAEF VGVSHLKQEP QKVKVSSSIQ HPETPLHISG FHLPYKPKPP QETEKGHSAC EPENLEFSSF MFWRNPLPNI DHELQELLID RGEDVPSEEE EEEENGFEDR KDDSDDDGGG WITPSNIKQI QQELEQCDVP EDVRVGCLTT DFAMQNVLLQ MGLHVLAVNG MLIREARSYI LRCHGCFKTT SDMSRVFCSH CGNKTLKKVS VTVSDDGTLH MHFSRNPKVL NPRGLRYSLP TPKGGKYAIN PHLTEDQRFP QLRLSQKARQ KTNVFAPDYI AGVSPFVEND ISSRSATLQV RDSTLGAGRR RLNPNASRKK FVKKR | |
Greater than 80.0% as determined by SDS-PAGE. |