Learn More
R&D Systems™ Recombinant Human Mcl-1 Western Blot Standard Protein
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Marke:  R&D Systems™ WBC003
Artikelnummer. 18335587
  • 221.00 CHF / 100 Mikroliter
Verfügbar ab: 29-01-2025
Sie sich ein, um den Lagerbestand zu sehen
Entdecken Sie weitere Sonderangebote
Beschreibung
The Recombinant Human Mcl-1 Western Blot Standard Protein has been validated for the following applications: Western Blot.
Spezifikation
Spezifikation
4170
100 μL
Unconjugated
Mcl-1 Western Blot Standard
Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Recombinant
Produktvorschläge