missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LZIC Protein
A cDNA sequence encoding the LZIC was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12611-1mg
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Spezifikation
LZIC Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSVKEGIEK AGVCPADNVR CFKSDPPQCH TDQDCLGERK CCYLHCGFKC VIPVKELEEG GNKDEDVSRP YPEPGWEAKC PGSSSTRCPQ K | |
Greater than 85% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The LZIC solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 1mM DTT and 10% glycerol. |