Learn More
enQuireBio™ Recombinant Human KLF4 Protein
A cDNA sequence encoding the KLF4 was constructed and used to recombinantly synthesize the protein.
141.00 CHF - 219.00 CHF
Spezifikation
Name | KLF4 Protein |
---|---|
Kennzeichnung | Research Use Only |
Endotoxin-Konzentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Produkttyp | Recombinant Protein |
Kreuzreaktivität | Human |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15980609
|
enQuireBio™
QP12498-HIS-5ug |
5 μg |
141.00 CHF
5 Mikrogramm |
Verfügbar ab: 05-03-2025
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15970609
|
enQuireBio™
QP12498-HIS-20ug |
20 μg |
219.00 CHF
20 Mikrogramm |
Verfügbar ab: 05-03-2025
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Spezifikation
KLF4 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
KLF4 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0) and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSMAVS DALLPSFSTF ASGPAGREKT LRQAGAPNNR WREELSHMKR LPPVLPGRPY DLAAATVATD LESGGAGAAC GGSNLAPLPR RETEEFNDLL DLDFILSNSL THPPESVAAT VSSSASASSS SSPSSSGPAS APSTCSFTYP IRAGNDPGVA PGGTGGGLLY GRESAPPPTA PFNLADINDV SPSGGFVAEL LRPELDPVYI PPQQPQPPGG GLMGKFVLKA SLSAPGSEYG SPSVISVSKG SPDGSHPVVV APYNGGPPRT CPKIKQEAVS SCTHLGAGPP LSNGHRPAAH DFPLGRQLPS RTTPTLGLEE VLSSRDCHPA LPLPPGFHPH PGPNYPSFLP DQMQPQVPPL HYQELMPPGS CMPEEPKPKR GRRSWPRKRT AT | |
Greater than 85% as determined by SDS-PAGE. |