Learn More
R&D Systems™ Recombinant Human Kallikrein 5 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity
Marke: R&D Systems™ 1108-SE-010
449.23 CHF gültig bis 2024-12-16
Benutzen Sie den Promotions-Code "21630" um Ihren Promotions-Preis zu erhalten.
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The Recombinant Human Kallikrein 5 Protein is derived from NS0. The Recombinant Human Kallikrein 5 Protein has been validated for the following applications: Enzyme Activity.Spezifikation
Supplied as a 0.2μm filtered solution in MES, NaCl and Glycerol. | |
31 kDa | |
10 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening. | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
25818 | |
Kallikrein 5 | |
Mouse myeloma cell line,NS0-derived human Kallikrein 5 protein Ile67-Ser293,with a C-terminal 10-His tag | |
Mature form | |
Recombinant |
Berichtigung von Produktinhalten