missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ITGB1 Protein
A cDNA sequence encoding the ITGB1 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP12462-HIS-20ug
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Spezifikation
ITGB1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MQTDENRCLK ANAKSCGECI QAGPNCGWCT NSTFLQEGMP TSARCDDLEA LKKKGCPPDD IENPRGSKDI KKNKNVTNRS KGTAEKLKPE DITQIQPQQL VLRLRSGEPQ TFTLKFKRAE DYPIDLYYLM DLSYSMKDDL ENVKSLGTDL MNEMRRITSDFRIGFGSFVE KTVMPYISTT PAKLRNPCTS EQNCTSPFSY KNVLSLTNKG EVFNELVGKQ RISGNLDSPE GGFDAIMQVA VCGSLIGWRN VTRLLVFSTD AGFHFAGDGK LGGIVLPNDG QCHLENNMYT MSHYYDYPSI AHLVQKLSEN NIQTIFAVTE EFQPVYKELK NLIPKSAVGT LSANSSNVIQ LIIDAYNSLS SEVILENGKL SEGVTISYKS YCKNGVNGTG ENGRKCSNIS IGDEVQFEIS ITSNKCPKKD SDSFKIRPLG FTEEVEVILQ YI | |
Greater than 80% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The ITGB1 solution (1 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol. |