missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ISG15 Protein

A cDNA sequence encoding the ISG15 was constructed and used to recombinantly synthesize the protein.

131.00 CHF - 2605.00 CHF

Spezifikation

Name ISG15 Protein
Kennzeichnung Research Use Only
Endotoxin-Konzentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Produkttyp Recombinant Protein
Kreuzreaktivität Human
Mehr Spezifikationen ansehen

Produkte 3
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
15970489
Unterlagen ansehen Aktionsdetails
enQuireBio™
QP12460-10ug
10 μg
131.00 CHF
10 Mikrogramm
Verfügbar ab: 11-12-2024
Sie sich ein, um den Lagerbestand zu sehen
Add to basket
 
15990489
missing translation for 'viewDocuments' Promotion Details
enQuireBio™
QP12460-50ug
50 μg
203.00 CHF
50 Mikrogramm
Verfügbar ab: 11-12-2024
Sie sich ein, um den Lagerbestand zu sehen
Add to basket
 
15980489
missing translation for 'viewDocuments' Promotion Details
enQuireBio™
QP12460-1mg
1 mg
2605.00 CHF
1 Milligramm
Verfügbar ab: 11-12-2024
Sie sich ein, um den Lagerbestand zu sehen
Add to basket
 
Description

Description

Specifications

Specifications

ISG15 Protein
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
Untagged
The ISG15 solution contains 20mM Tris-HCl pH 8.0, 100mM NaCl and 1mM DTT.
Research Use Only
Recombinant Protein
E. coli
MADQLTEEQI AEFKEAFSLF DKDGDGTITT KELGTVMRSL GQNPTEAELQ DMINEVDADG NGTIDFPEFL TMMARKMKDT DSEEEIREAF RVFDKDGNGY ISAAELRHVM TNLGEKLTDE EVDEMIREAD IDGDGQVNYE EFVQMMTAKG SMGWDLTVKM LAGNEFQVSL SSSMSVSELK AQITQKIGVH AFQQRLAVHP SGVALQDRVP LASQGLGPGS TVLLVVDKCD EPLNILVRNN KGRSSTYEVR LTQTVAHLKQ QVSGLEGVQD DLFWLTFEGK PLEDQLPLGE YGLKPLSTVF MNLRLRGG
Greater than 98.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Videos
SDS
Documents

Documents

Certificates