missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HERPUD1 Protein
A cDNA sequence encoding the HERPUD1 was constructed and used to recombinantly synthesize the protein.
131.00 CHF - 7525.00 CHF
Spezifikation
Name | HERPUD1 Protein |
---|---|
Kennzeichnung | Research Use Only |
Endotoxin-Konzentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Produkttyp | Recombinant Protein |
Kreuzreaktivität | Human |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15989728
|
enQuireBio™
QP12218-2ug |
2 μg |
131.00 CHF
2 Mikrogramm |
Verfügbar ab: 29-10-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15969728
|
enQuireBio™
QP12218-10ug |
10 μg |
203.00 CHF
10 Mikrogramm |
Verfügbar ab: 29-10-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15979728
|
enQuireBio™
QP12218-1mg |
1 mg |
7525.00 CHF
1 Milligramm |
Verfügbar ab: 29-10-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Spezifikation
HERPUD1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The HERPUD1 solution (0.5 mg/1ml) contains Phosphate buffer saline (pH 7.4) and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMESETEP EPVTLLVKSP NQRHRDLELS GDRGWSVGHL KAHLSRVYPE RPRPEDQRLI YSGKLLLDHQ CLRDLLPKQE KRHVLHLVCN VKSPSKMPEI NAKVAESTEE PAGSNRGQYP EDSSSDGLRQ REVLRNLSSP GWENISRPEA AQQAFQGLGP GFSGYTPYGW LQLSWFQQIY ARQYYMQYLA ATAASGAFVP PPSAQEIPVV SAPAPAPIHN QFPAENQPAN QNAAPQVVVN PGANQNLRMN AQGGPIVEED DEINRD | |
Greater than 85.0% as determined by SDS-PAGE. |