missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GMP reductase 2 Protein
A DNA sequence encoding the Homo sapiens (Human) GMP reductase 2, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP8126-ec-10ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
NP_001002000.1 | |
GMP reductase 2 Protein | |
Research Use Only | |
GMPR2 | |
Recombinant Protein | |
E. coli | |
MTSCLPALRF IATPRLSAMP HIDNDVKLDF KDVLLRPKRS TLKSRSEVDL TRSFSFRNSK QTYSGVPIIA ANMDTVGTFE MAKVLCKFSL FTAVHKHYSL VQWQEFAGQN PDCLEHLAAS SGTGSSDFEQ LEQILEAIPQ VKYICLDVAN GYSEHFVEFV KDVRKRFPQH TIMAGNVVTG EMVEELILSG ADIIKVGIGP GSVCTTRKKT GVGYPQLSAV MECADAAHGL KGHIISDGGC SCPGDVAKAF GAGADFVMLG GMLAGHSESG GELIERDGKK YKLFYGMSSE MAMKKYAGGV AEYRASEGKT VEVPFKGDVE HTIRDILGGI RSTCTYVGAA KLKELSRRTT FIRVTQQVNP IFSEAC | |
Greater than 90% as determined by SDS-PAGE. |
51292 | |
10 μg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |