missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human FGF acidic/FGF1 (aa 2-155) Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Marke: R&D Systems™ 231-BC-025
316.19 CHF gültig bis 2024-12-16
Benutzen Sie den Promotions-Code "21630" um Ihren Promotions-Preis zu erhalten.
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The Recombinant Human FGF acidic (aa 2-155) Protein is derived from E. coli. The Recombinant Human FGF acidic (aa 2-155) Protein has been validated for the following applications: Bioactivity.Spezifikation
Lyophilized from a 0.2μm filtered solution in MOPS, Na2SO4, TCEP and EDTA with BSA as a carrier protein. | |
17 kDa | |
25 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 3 months, ≤ -20°C under sterile conditions after reconstitution. | |
Unconjugated | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
2246 | |
FGF acidic aa 2-155 | |
E. coli-derived human FGF acidic/FGF1 protein Ala2-Asp155 | |
AFGF, alpha, alpha-ECGF, beta-ECGF, ECGF, ECGFB, ECGF-betaAcidic fibroblast growth factor, endothelial cell growth factor, beta, FGF acidic, FGF-1, FGFABeta-endothelial cell growth factor, FGF-alpha, fibroblast growth factor 1 (acidic), GLIO703, HBGF1, HBGF-1, heparin-binding growth factor 1 | |
Recombinant |