Learn More
enQuireBio™ Recombinant Human Fatty acid desaturase 2 Protein
A DNA sequence encoding the Homo sapiens (Human) Fatty acid desaturase 2, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP6016-ec-50ug
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Spezifikation
NP_001268430.1 | |
Fatty acid desaturase 2 Protein | |
Research Use Only | |
FADS2 | |
Recombinant Protein | |
E. coli | |
MGKGGNQGEG AAEREVSVPT FSWEEIQKHN LRTDRWLVID RKVYNITKWS IQHPGGQRVI GHYAGEDATD AFRAFHPDLE FVGKFLKPLL IGELAPEEPS QDHGKNSKIT EDFRALRKTA EDMNLFKTNH V | |
Greater than 90% as determined by SDS-PAGE. |
9415 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |