missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Cyclic GMP-AMP synthase Protein
A DNA sequence encoding the Homo sapiens (Human) Cyclic GMP-AMP synthase, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP7853-ec-200ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
NP_612450.2 | |
Cyclic GMP-AMP synthase Protein | |
Research Use Only | |
MB21D1 | |
Recombinant Protein | |
E. coli | |
GASKLRAVLE KLKLSRDDIS TAAGMVKGVV DHLLLRLKCD SAFRGVGLLN TGSYYEHVKI SAPNEFDVMF KLEVPRIQLE EYSNTRAYYF VKFKRNPKEN PLSQFLEGEI LSASKMLSKF RKIIKEEIND IKDTDVIMKR KRGGSPAVTL LISEKISVDI TLALESKSSW PASTQEGLRI QNWLSAKVRK QLRLKPFYLV PKHAKEGNGF QEETWRLSFS HIEKEILNNH GKSKTCCENK EEKCCRKDCL KLMKYLLEQL KERFKDKKHL DKFSSYHVKT AFFHVCTQNP QDSQWDRKDL GLCFDNCVTY FLQCLRTEKL ENYFIPEFNL FSSNLIDKRS KEFLTKQIEY ERNNEFPVFD EF | |
Greater than 90% as determined by SDS-PAGE. |
115004 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |