Learn More
R&D Systems™ Recombinant Human Complement Factor H-related 5/CFHR5
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity
Marke:  R&D Systems™ 3845-F5-050
466.51 CHF gültig bis 2024-12-16
Benutzen Sie den Promotions-Code "21630" um Ihren Promotions-Preis zu erhalten.
Artikelnummer. 18136820
  • 600.00 CHF / 50 Mikrogramm
Verfügbar ab: 22-01-2025
Sie sich ein, um den Lagerbestand zu sehen
Entdecken Sie weitere Sonderangebote
Beschreibung
The Recombinant Human Complement Factor H-related 5/CFHR5 is derived from NS0. The Recombinant Human Complement Factor H-related 5/CFHR5 has been validated for the following applications: Binding Activity.
Spezifikation
Spezifikation
Lyophilized from a 0.2μm filtered solution in Tris and NaCl.
64 kDa
50 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant
81494
Complement Factor H-related 5/CFHR5
Mouse myeloma cell line,NS0-derived human Complement Factor H-related 5/CFHR5 protein Glu19-Glu569,with a C-terminal 10-His tag
Unconjugated
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.