missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Cdc42 / Rac Protein
A DNA sequence encoding the Homo sapiens (Human) Cdc42/Rac, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP10060-ba-100ug
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Spezifikation
NP_065074.1 | |
Cdc42 / Rac Protein | |
Research Use Only | |
PAK7 | |
Recombinant Protein | |
Baculovirus | |
MFGKKKKKIE ISGPSNFEHR VHTGFDAQEQ KFTGLPQQWH SLLADTANRP KPMVDPSCIT PIQLAPMKTI VRGNKPCKET SINGLLEDFD NISVTRSNSL RKESPPTPDQ GASSHGPGHA EENGFITFSQ YSSESDTTAD YTTEKYREKS LYGDDLDPYY RGSHAAKQNG HVMKMKHGEA YYSEVKPLKS DFARFSADYH SHLDSLSKPS EYSDLKWEYQ RASSSSPLDY SFQFTPSRTA GTSGCSKESL AYSESEWGPS LDDYDRRPKS SYLNQTSPQP TMRQRSRSGS GLQ | |
Greater than 90% as determined by SDS-PAGE. |
57144 | |
100 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |