missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human CCL23/Ck beta 8-1 (aa 46-137) Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Marke:  R&D Systems™ 508-CK-025

272.99 CHF gültig bis 2024-12-16
Benutzen Sie den Promotions-Code "21630" um Ihren Promotions-Preis zu erhalten.


 Lernen Sie weitere Varianten dieses

Artikelnummer. 18360318

  • 349.00 CHF / 25 Mikrogramm
Verfügbar ab: 20-01-2025
Sie sich ein, um den Lagerbestand zu sehen
Entdecken Sie weitere Sonderangebote
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Beschreibung

Beschreibung

The Recombinant Human CCL23/Ck beta 8-1 (aa 46-137) Protein is derived from E. coli. The Recombinant Human CCL23/Ck beta 8-1 (aa 46-137) Protein has been validated for the following applications: Bioactivity.
Spezifikation

Spezifikation

Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
10.5 kDa
25 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unconjugated
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
6368
CCL23/Ck beta 8-1 aa 46-137
E. coli-derived human CCL23/Ck beta 8-1 protein Met46-Asn137
C6 beta-chemokine, chemokine (C-C motif) ligand 23, Ck beta 8-1, Ckb-8, CKb8, Ckb81, CK-beta-8, hmrp-2a, Macrophage inflammatory protein 3, MIP3, MIP-3CK-BETA-8, MPIF1, MPIF-1Ckb-8-1, Myeloid progenitor inhibitory factor 1, SCYA23C-C motif chemokine 23, small inducible cytokine subfamily A (Cys-Cys), member 23, Small-inducible cytokine A23
Recombinant
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote