missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human alpha-Galactosidase A/GLA Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Marke: R&D Systems™ 6146-GH-020
423.31 CHF gültig bis 2024-12-16
Benutzen Sie den Promotions-Code "21630" um Ihren Promotions-Preis zu erhalten.
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Measured by its ability to hydrolyze 4-methylumbelliferyl-alpha-D-galactopyranoside. The specific activity is >1,100 pmol/min/μg, as measured under the described conditions.Spezifikation
P06280 | |
Supplied as a 0.2 μm filtered solution in Tris and NaCl. | |
M.W. (Observed): 45-55 kDa, reducing conditions; M.W. (theoretical): 46 kDa | |
20 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening. | |
GLA | |
Recombinant | |
alpha-Galactosidase A/GLA |
Enzyme Activity | |
2717 | |
alpha-Galactosidase A/GLA | |
Chinese Hamster Ovary cell line, CHO-derived human alpha-Galactosidase A/GLA protein Met1-Leu429, with a C-terminal 6-His tag | |
<1.0 EU / 1 μg of the protein by the LAL method. | |
Unconjugated | |
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining. |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur