missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Cynomolgus HGFR/c-MET Fc Chimera Protein
Marke: R&D Systems™ 11267-ME-100
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Measured by its binding ability in a functional ELISA. When Recombinant Human HGF (NS0-expressed) Protein (Catalog # 294-HGN ) is immobilized at 0.250 μg/mL (100 μL/well), Recombinant Cynomolgus Monkey HGFR/c-MET Fc Chimera (Catalog # 11267-ME) binds with an ED50 of 2.50-25.0 ng/mL.Spezifikation
EHH52447.1 | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. | |
100 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. | |
Unconjugated | |
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining. |
Bioactivity | |
M.W.-observed: 135-150 kDa, 40-45 kDa, and 103-115 kDa, under reducing conditions., M.W.-theroretical: 128 kDa, 33 kDa (alpha chain), and 96 kDa (beta chain) | |
Human embryonic kidney cell, HEK293-derived cynomolgus monkey HGFR/c-MET protein Cynomolgus HGFR/c-MET (Glu25-Thr932) (C-terminus) Accession # EHH52447.1 IEGRMD Human IgG1 (Pro100-Lys330) (N-terminus) | |
<0.10 EU / 1 μg of the protein by the LAL method. | |
Reconstitute at 500 μg/mL in PBS. |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur