missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Canine IL-10 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Marke: R&D Systems™ 735-CL-010/CF
377.53 CHF gültig bis 2024-12-16
Benutzen Sie den Promotions-Code "21630" um Ihren Promotions-Preis zu erhalten.
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The Recombinant Canine IL-10 Protein is derived from E. coli. The Recombinant Canine IL-10 Protein has been validated for the following applications: Bioactivity.Spezifikation
Lyophilized from a 0.2μm filtered solution in NaH2PO4 and NaCl. | |
18.9 kDa | |
10μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
CSIF, CSIFMGC126450, Cytokine synthesis inhibitory factor, GVHDS, IL10, IL10A, IL-10MGC126451, interleukin 10, interleukin-10, TGIF | |
Recombinant |
403628 | |
IL-10 | |
E. coli-derived canine IL-10 protein Ser20-Ile179 | |
Noncovalently-linked homodimer | |
Unconjugated | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |