missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Rat P-Selectin ELISA Kit
Rat P-Selectin quantitates rat P-Selectin in serum, plasma, supernatant.
836.00 CHF - 6320.00 CHF
Spezifikation
Zugriffsnummer | CD62; CD62 antigenähnliches Familienmitglied P; CD62P; CD62P-Antigen; CD62P; CD62-Homolog; FLJ45155; GMP140; GMP-140; GMRP; Granulat-Membranprotein 140; Granulat-Membranprotein 140 kDa; Granulozyten-Membranprotein; GRMP; LECAM3; Leukozyten-endotheliales Zelladhäsionsmolekül 3; PADGEM; Thrombozyten-Aktivierung-abhängiges Granulat-Außenmembranprotein; Thrombozyten-Alpha-Granulat-Membranprotein; PSEL; PSELECT; P-Selektin; P-Selektin-Vorläufer; RP1-86F14.2; Selektin P; Selektin P (Granulatmembranprotein 140 kDa, Antigen CD62); Selektin, Thrombozyten; Selp |
---|---|
Assay-Bereich | 40 bis 10.000 pg/ml |
Assay-Empfindlichkeit: | 40 pg/mL |
Konjugat | Biotin |
Produkttyp | ELISA |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
17165889
|
Invitrogen™
ERA51RBX5 |
5 x 96 Tests |
2895.00 CHF
1 Stück |
Verfügbar ab: 02-08-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
17165063
|
Invitrogen™
ERA51RB |
96 Tests |
836.00 CHF
96 Stück |
Verfügbar ab: 05-08-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
18577934
|
Invitrogen™
ERA51RBX10 |
10 x 96 Tests |
6320.00 CHF
10 Stück |
Verfügbar ab: 05-08-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
Rat P-Selectin quantitates rat P-Selectin in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant rat P-Selectin.
P-selectin (Selectin P, GMP-140, LECAM3, CD62 antigen-like family member) is a 140 kDa type 1 transmembrane glycoprotein, and is one of the most commonly studied proteins that regulate cell rolling. P-selectin is stored in the Weibel-Palade bodies of endothelial cells, as well as in a-granules of platelets. From there, it can be rapidly brought to the cell surface after exposure to thrombin, histamine, complement 5a, Ca21 ionophores, or adenosine diphosphate. P-selectin protein redistributes to the plasma membrane during platelet activation and degranulation and mediates the interaction of activated endothelial cells or platelets with leukocytes. P-selectin plays an important role in adhesive processes between leucocytes and endothelial cells, and is a calcium dependent receptor that binds to sialylated forms of Lewis blood group carbohydrate antigens found on neutrophils and monocytes. P-selectin is constitutively expressed in inflammation and contributes to atherogenesis, thrombosis and tissue destruction. Clinically, P-selectin is used to distinguish heparin induced thrombocytopenia with and without thrombosis.Spezifikation
CD62; CD62 antigenähnliches Familienmitglied P; CD62P; CD62P-Antigen; CD62P; CD62-Homolog; FLJ45155; GMP140; GMP-140; GMRP; Granulat-Membranprotein 140; Granulat-Membranprotein 140 kDa; Granulozyten-Membranprotein; GRMP; LECAM3; Leukozyten-endotheliales Zelladhäsionsmolekül 3; PADGEM; Thrombozyten-Aktivierung-abhängiges Granulat-Außenmembranprotein; Thrombozyten-Alpha-Granulat-Membranprotein; PSEL; PSELECT; P-Selektin; P-Selektin-Vorläufer; RP1-86F14.2; Selektin P; Selektin P (Granulatmembranprotein 140 kDa, Antigen CD62); Selektin, Thrombozyten; Selp | |
40 pg/mL | |
ELISA | |
P98106. | |
<12% | |
Vorbeschichtete 96-Mikrotiterplatte, Standard, Assay-Verdünnungskonzentrat, biotinylierter Nachweisantikörper, SAV-HRP, Waschpuffer, Chromogen, Stopplösung, Dichtungsfolien für Mikrotiterplatten | |
Nur für Forschungszwecke | |
P-Selektin (CD62P) | |
4 h 45 min |
40 bis 10.000 pg/ml | |
Biotin | |
Kolorimetrisches Mikrotiterplatten-Lesegerät | |
SELP | |
<10% | |
HRP | |
2 bis 8 °C | |
1 h 20 min |