Learn More
Novus Biologicals™ RAI2 Overexpression Lysate
Marke: Novus Biologicals™ NBL1-15129
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The RAI2 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.Spezifikation
The lysate was created in HEK293T cells, using plasmid ID RC26638 and based on accession number NM_21785. The protein contains a C-terminal DDK tag. | |
0.1 mg | |
Protein | |
-80°C. Avoid freeze-thaw cycles. |
Western Blot | |
RIPA buffer | |
Homo sapiens retinoic acid induced 2 (RAI2), mRNA. | |
Native |
Nur für Forschungszwecke