Learn More
Novus Biologicals™ RAI2 Overexpression Lysate
Marke:  Novus Biologicals™ NBL1-15129
Artikelnummer. 18109579
  • 820.00 CHF / 0.10 Milligramm
Verfügbar ab: 20-12-2024
Sie sich ein, um den Lagerbestand zu sehen
Entdecken Sie weitere Sonderangebote
Beschreibung
The RAI2 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
Spezifikation
Spezifikation
The lysate was created in HEK293T cells, using plasmid ID RC26638 and based on accession number NM_21785. The protein contains a C-terminal DDK tag.
0.1 mg
Protein
-80°C. Avoid freeze-thaw cycles.
Western Blot
RIPA buffer
Homo sapiens retinoic acid induced 2 (RAI2), mRNA.
Native
Sonderangebote
Nur für Forschungszwecke