missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Guinea Pig IgG (H+L) Secondary Antibody, FITC, Invitrogen™
Rabbit Polyclonal Secondary Antibody
Marke: Invitrogen PA128593
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Anti-Guinea Pig secondary antibodies are affinity-purified antibodies with well-characterized specificity for guinea pig immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Spezifikation
Guinea Pig IgG (H+L) | |
Polyclonal | |
FITC | |
Rabbit | |
IgG fraction | |
RUO | |
Guinea Pig | |
Liquid |
Flow Cytometry, Immunocytochemistry | |
2.2 mg/mL | |
0.01M sodium phosphate with 0.15M NaCl, 10mg/mL BSA and 0.01% thimerosal; pH 7.2 | |
guinea pig whole molecule | |
1 mg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
IgG |