Learn More
R&D Systems™ Mouse ChemR23 (NP_032179) VersaClone cDNA
Chemokine-like Receptor 1/Orphan G Protein-coupled Receptor
Marke: R&D Systems™ RDC0225
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The Mouse ChemR23 (NP_032179) VersaClone cDNA clone contains the complete ORF for ChemR23/CMKLR1, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The ChemR23/CMKLR1 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.Bestellinformation
Ships at ambient temperature.
Spezifikation
mChemR23 | |
10 μg at 0.2 μg/μL | |
1128bp | |
Maus |
NP_032179 | |
cDNA is provided in 10mM Tris-Cl, pH 8.5 | |
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
For Reasearch Use Only; Not for Use in Humans