missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse Aminoacylase/ACY1 Protein, CF
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Marke: R&D Systems™ 2700-ZN-020
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Measured by its ability to cleave N-acetyl-L-Methione (Ac-Met). The specific activity is >4,000 pmol/min/μg, as measured under the described protocol.Spezifikation
NP_079647 | |
Supplied as a 0.2 μm filtered solution in Tris and NaCl. | |
M.W. (Observed): 42 kDa, reducing conditions; M.W. (theoretical): 45 kDa | |
20 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening. | |
Acy1 | |
Recombinant | |
Aminoacylase/ACY1 |
Enzyme Activity | |
95 | |
Aminoacylase/ACY1 | |
Mouse myeloma cell line, NS0-derived mouse Aminoacylase/ACY1 protein Met1-Ser408, with a C-terminal 10-His tag Accession # NP_079647 | |
<1.0 EU / 1 μg of the protein by the LAL method. | |
Unconjugated | |
>95%, by SDS-PAGE under reducing conditions and visualized by silver stain |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur