Learn More
R&D Systems™ Human VAChT/SLC18A3 (AAA20497) VersaClone cDNA
Vesicular Acetylcholine Transporter/Solute Carrier Family 18 Member 3
Marke: R&D Systems™ RDC0901
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The Human VAChT/SLC18A3 (AAA20497) VersaClone cDNA clone contains the complete ORF for VAChT/SLC18A3, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The VAChT/SLC18A3 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.Bestellinformation
Ships at ambient temperature.
Spezifikation
hSLC18A3 | |
10 μg at 0.2 μg/μL | |
1611bp | |
Human |
AAA20497 | |
cDNA is provided in 10mM Tris-Cl, pH 8.5 | |
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. | |
10 μg |
Berichtigung von Produktinhalten
Name des Produkts
For Reasearch Use Only; Not for Use in Humans
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur