Learn More
R&D Systems™ Human Leukotriene B4 R1 (NP_858043) VersaClone cDNA
Leukotriene B4 Receptor
Marke:  R&D Systems™ RDC0413
Artikelnummer. 18461620
  • 328.00 CHF / 10 Mikrogramm
Verfügbar ab: 20-12-2024
Sie sich ein, um den Lagerbestand zu sehen
Entdecken Sie weitere Sonderangebote
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
The Human Leukotriene B4 R1 (NP_858043) VersaClone cDNA clone contains the complete ORF for Leukotriene B4 R1, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The Leukotriene B4 R1 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Bestellinformation
Ships at ambient temperature.
Spezifikation
Spezifikation
hLTB4R
10 μg at 0.2 μg/μL
1072bp
Human
NP_858043
cDNA is provided in 10mM Tris-Cl, pH 8.5
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Sonderangebote
Sonderangebote
For Reasearch Use Only; Not for Use in Humans