missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Holo-Transferrin Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
193.00 CHF - 804.00 CHF
Spezifikation
Zusammensetzung | Lyophilized from a 0.2 μm filtered solution in NH 4 HCO 3 . |
---|---|
Gen-ID (Entrez) | 7018 |
Name | Holo-Transferrin |
Quelle | Human plasma-derived Holo-Transferrin protein The human plasma used for the isolation of this product were certified by the supplier to be HIV-1 and HBsAg negative at the time of shipment. Human blood products should always be treated in accordance with universal handling precautions. |
Lagerungsbedingungen | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
18108290
|
R&D Systems™
2914-HT-100MG |
100 mg |
193.00 CHF
100 Milligramm |
Verfügbar ab: 10-01-2025
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
18361698
|
R&D Systems™
2914-HT-001G |
1 g |
804.00 CHF
1 Gramm |
Verfügbar ab: 10-01-2025
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
The Human Holo-Transferrin Protein is derived from Human Plasma. The Human Holo-Transferrin Protein has been validated for the following applications: Bioactivity.Spezifikation
Lyophilized from a 0.2 μm filtered solution in NH 4 HCO 3 . | |
Holo-Transferrin | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
90%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
7018 | |
Human plasma-derived Holo-Transferrin protein The human plasma used for the isolation of this product were certified by the supplier to be HIV-1 and HBsAg negative at the time of shipment. Human blood products should always be treated in accordance with universal handling precautions. | |
Unconjugated |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts