missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Bacterial Thermolysin Protein, CF
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity
Marke: R&D Systems™ 3097-ZN-020
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Measured by its ability to cleave the fluorogenic peptide substrate, Mca-RPPGFSAFK(Dnp)-OH (Catalog # ES005 ). The specific activity is >12,000 pmol/min/μg, as measured with 10 μM ES005 and 2 ng enzyme in 100 μL of 50 mM Tris, 10 mM CaCl2, 0.15 M NaCl, 0.05% Brij 35, pH 7.5.Spezifikation
Enzyme Activity | |
M.W. (Observed): 36 kDa, reducing conditions | |
20 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening. | |
Unconjugated | |
Thermolysin |
Supplied as a 0.2 μm filtered solution in Tris, NaCl, CaCl2 and Glycerol. | |
Thermolysin | |
Geobacillus stearothermophilus -derived Thermolysin protein | |
<1.0 EU / 1 μg of the protein by the LAL method. | |
>70%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining. |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur