Learn More
Thermo Scientific™ Quartz tubing, fused, 19mm (0.748in) OD, 16mm (0.630in) ID
Manufactured by fusing naturally occurring crystalline silica
Marke:  Thermo Scientific™ 043645.NT
Artikelnummer. 11325867
  • 182.57 CHF /
Verfügbar ab: 19-12-2024
Sie sich ein, um den Lagerbestand zu sehen
Entdecken Sie weitere Sonderangebote
Beschreibung
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
Fused Quartz
0.748 in.
19 mm
122 cm