missing translation for 'onlineSavingsMsg'
Learn More

Melford Proteinase K

From Tritirachium album.

Marke:  Melford P50220-0.5

 Lernen Sie weitere Varianten dieses

Artikelnummer. 30018716

  • 708.00 CHF / 500 Milligramm
Verfügbar ab: 18-12-2024
Sie sich ein, um den Lagerbestand zu sehen
Entdecken Sie weitere Sonderangebote
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Beschreibung

Beschreibung

From Tritirachium album.

From Tritirachium album. Serine protease for proteolytic inactivation of nucleases during isolation of DNA and RNA.

Description

Proteinase K is a highly reactive serine protease that displays an ability to digest native proteins, thereby inactivating enzymes such as DNase and RNase without recourse to a denaturation process. It is the most powerful proteinase among all proteinases characterized so far. It cleaves at the peptide bond adjacent to the carboxylic acid group of aliphatic, aromatic, or hydrophobic amino acids.

Recombinant Proteinase K is used in the isolation or preparation of high molecular weight nucleic acids. It is highly pure has a higher specific activity and is more stable at room temperature when compared to native Proteinase K.

Recombinant Proteinase K is expressed from yeast Pichia pastoris with a cloned gene encoding Engyodontium album (Tritirachium album) endolytic protease.

Applications

  • Recombinant Proteinase K is extremely effective on native proteins and can therefore be used to rapidly inactivate endogenous nucleases such as RNases and DNases. This property makes Proteinase K particularly suitable for the isolation of native RNA and DNA from tissues or cell lines.
  • Used for the analysis of membrane structures by modifying proteins and glycoproteins on cell surfaces.
  • Particularly well suited for isolating nucleic acids for amplification reactions.
  • Promotes cell lysis by activating a bacterial autolytic factor.
  • Used to remove cellular debris during the preparation of colony lifts, and to treat tissue sections to ensure efficient probe infiltration during in situ hybridization.

Reconstitution Recommendations:

For stock solution preparation, we suggest reconstituting the lyophilized powder in a buffer (20 mM Tris, 1 mM CaCl2, 2% Glycerol, pH 7.4).

Calcium is important for thermo-stability of Proteinase K and pH value is critical for high solubility.

The use of cryoprotectant glycerol to a final concentration of 50% in stock solution is to prevent freeze/thaw damage. Proteinase K retained 99% of activity after 12 freeze/thaw cycles in stock solution. Avoid repeated freezing and thawing to prevent precipitation of the protein if glycerol is not added.

Proteinase K solution remains active over a broad pH range (4.0-12.5, often used in range of 7.5-9.0) and over a broad temperature range of 25°C to 75°C. Solution remains sterile and stable for at least 12 months at 4°C.

Caution: Proteinase K Recombinant Lyophilized Powder is not sterile. Stock solution can be prepared as a 20-40 mg/mL in 20 mM Tris-HCl buffer, sterilized using a 0.22 μm filter. Stock solution can be stored in 50% glycerol in aliquots at a wide temperature range of 24°C to -80°C.

For sterile filtration, we recommend using PES pr PVDF membranes with low protein binding.

Composition (NO ADDITIVES):

  • ≥90% Protein Proteinase K, biuret test.
  • Excipients: Sucrose 8%
  • DNA and RNA free
  • Endotoxin Free

Specifications:

  • Stable and active over a wide pH range of 4 to 12, Maximal Activity is in pH range of 7.5 to 12.0
  • Active with SDS, urea and EDTA. The most active temperature is 65°C.
  • Inactivated by diisopropyl fluorophosphates (DFP) and phenyl methane sulfonyl fluoride (PMSF).
  • Purification: Nickel affinity chromatography followed by ion-exchange chromatography and dialysis.
  • EC : 3.4.21.64
Spezifikation

Spezifikation

39450-01-6
500 mg
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote

Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt